Learn More
Justman Brush Brush with Extension for Flask Curves
Shop All Fisherbrand Products
Click to view available options
Color:
White
Unit Size:
Each
Tekniske data
Tekniske data
| Product Type | Flask Brush |
| Bristle Material | Unbleached Hog Bristle |
| Handle Material | Plastic |
| Color | White |
| Diameter (English) Bristle | 1.5 in. |
| Diameter (Metric) Bristle | 3.81 cm |
| For Use With (Application) | For cleaning bottom curves and inside shoulders of flasks and bottles |
| Length (English) | 16 in. |
| Length (English) Bristle | 4 in. |
| Length (Metric) | 40.64 cm |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion