Learn More
Fisherbrand™ Heavy-Duty/Utility Funnels
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
40 mL
100 mL
140 mL
210 mL
410 mL
500 mL
Diameter (Metric)Top:
6.5cm
8.7cm
10.3cm
12.5cm
15.5cm
20.3 cm
Height (Metric):
9 cm
11 cm
11.5 cm
14.1 cm
17.7 cm
20.5 cm
Unit Size:
Pack of 12
Pack of 24
Pack of 3
Pack of 72
Specifications
Specifications
| Diameter (Metric)Top | 12.5cm |
| Capacity (Metric) | 210 mL |
| Material | Polypropylene |
| Height (Metric) | 14.1 cm |
| Autoclavable | Autoclavable |