Learn More
Fisherbrand™ Hygrometer Replacement Battery
Product Code. 11735495 Shop All Fisherbrand Products
Click to view available options
Battery Type:
357
392
AAA
Product Type:
Button Cell Battery
Cylindrical Battery
Voltage:
1.5 V
Unit Size:
Each
Product Code. 11735495
Hygrometer Replacement Battery for use with Hygrometers.
  • Come with lithium, button cell and 'AAA' alkaline battery
  • The electrical requirements are 3 V, 1.5 V and 1.5 V respectively
Specifications
Product Type Button Cell Battery
Battery Type 392
Material Alkaline
Cell Type Button
For Use With (Equipment) For hygrometers
Voltage 1.5 V