Learn More
Fisherbrand™ Multipurpose Specimen Storage Containers
Product Code. 11709388 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
118 mL
148 mL
237 mL
473 mL
946 mL
1100 mL
1900 mL
2500 mL
5100 mL
Closure Material:
LDPE
Polypropylene
Color:
Translucent
White
Shape:
Short, Wide
Tall, Thin
Unit Size:
Pack of 10
Pack of 100
Pack of 25
Pack of 300
Pack of 50
Product Code. 11709388
Inert to most chemicals including formaldehyde, weak acids and all bases
Fisherbrand™ Multipurpose Specimen Containers are designed for collection, transport, and storage of liquid and dry specimens and samples; nonsterile. Inert to most chemicals including formaldehyde, weak acids and all bases.
  • Resist freezing and boiling
  • Stackable for compact storage
  • Select containers are autoclavable (see ordering table); lids are not autoclavable
  1. Short/wide style
  2. Tall/thin style
  3. Graduated
Specifications
Product Type Specimen Container
Material Polypropylene
Shape Tall, Thin
Capacity (English) 5.0 oz.
Capacity (Metric) 148 mL
Closure Material Polypropylene
Color Translucent
Diameter (Metric) 5.78 cm
Height (English) 2.65 in.
Height (Metric) 6.73 cm
No. per Pack 300/Pk.