Learn More
Fisherbrand™ Polypropylene Graduated Specimen Containers
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
133 mL
148 mL
237 mL
Capacity (English):
4.5 oz.
5 oz.
8 oz.
Sterility:
Non-sterile
Sterile
Closure Included:
No
Yes
Unit Size:
Pack of 100
Pack of 500
Specifications
Specifications
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Sterility | Sterile |
| Capacity (English) | 5 oz. |
| Capacity (Metric) | 148 mL |