missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polyvinyl-Lined Polypropylene Caps

Product Code. 15522331
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Color White
Description Screw cap
Material HDPE
No. per Case 12/Pk.
Quantity 288/PK.
Liner Material Polyvinyl
Neck Finish 70-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner Polyvinyl
Closure Material Plastic
Closure Size 70-400 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'