Learn More
Fisherbrand™ Powder Funnels
GREENER_CHOICE
Product Code. 11997904 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric)Top:
6.5cm
7.5cm
10.5cm
15cm
Diameter (Metric) Stem:
16mm
21mm
27mm
Capacity (Metric):
50 mL
100 mL
225 mL
750 mL
Height (Metric):
6.6 cm
8.4 cm
10.3 cm
13.7 cm
Unit Size:
Pack of 12
Pack of 24
Pack of 36
Pack of 4
Pack of 6
Product Code. 11997904
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Parallel stems minimize bridging of powder.
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 10.5cm
Capacity (Metric) 225 mL
Diameter (Metric) Stem 21mm
Material Polypropylene
Height (Metric) 10.3 cm
Autoclavable Autoclavable
Length (Metric) Stem 33mm