Learn More
Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes
Product Code. 12711675
Click to view available options
Path Length:
1 mm
10 mm
Material:
Quartz Suprasil 300
Silica
Quantity:
2/Pk.
Ea.
Unit Size:
Each
Product Code. 12711675
Brand: Hellma™ 6030UV1053E
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Fitted PTFE covers or tapered PTFE stoppers, except 14-385-906A and -902A, which come with glass cover plates
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes provide optically excellent transparency.
  • Quartz - high UV transmission and low thermal expansion
  • Glass - used in the visible range of the spectrum
  • Low values of refractive index and dispertion
  • Good resistance to chemicals
Specifications
Material Silica
Quantity 2/Pk.
Path Length 10 mm