Learn More
Fisherbrand™ Stainless Steel Bow Tongs
Designed using high quality components these corrosion resistant, non-magnetic bow tongs feature excellent heat resistance and great cryogenic stability.
Brand:  Fisherbrand™ 3160
 View more versions of this product
Product Code. 11543522
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • 304 spec 18/8 non-magnetic stainless steel laboratory bow tongs with PTFE coated jaws
  • Corrosion resistant, non-magnetic bow tongs
  • Applied as a self priming single coat PTFE
  • Excellent heat resistance and great cryogenic stability
  • Excellent non-wetting properties that makes cleaning very easy and can also undergo steam or chemical sterilization
  • The coefficient of friction is generally in the range of 0.05 to 0.20
Specifications
Specifications
1 Ea.
Bow
Stainless Steel
PTFE
150 mm
Tongs