Learn More
Fisherbrand™ Stainless Steel Bow Tongs
Designed using high quality components these corrosion resistant, non-magnetic bow tongs feature excellent heat resistance and great cryogenic stability.
Brand: Fisherbrand™ 3160
Description
- 304 spec 18/8 non-magnetic stainless steel laboratory bow tongs with PTFE coated jaws
- Corrosion resistant, non-magnetic bow tongs
- Applied as a self priming single coat PTFE
- Excellent heat resistance and great cryogenic stability
- Excellent non-wetting properties that makes cleaning very easy and can also undergo steam or chemical sterilization
- The coefficient of friction is generally in the range of 0.05 to 0.20
Specifications
1 Ea. | |
Bow | |
Stainless Steel |
PTFE | |
150 mm | |
Tongs |