Learn More
SCAT™ Fitting with Ferrule for HPLC Systems
Shop All SCAT Europe Products
Click to view available options
Diameter (Metric) Outer:
1.6 mm
3.2 mm
Color:
Black
Colorless
White
Unit Size:
Pack of 5
Description
SCAT™ fittings are available in the following versions:
- PFA fitting with integrated ferrule in various colors and diameters
- Tube connectors in various diameters
Specifications
Specifications
| Type | Fitting |
| Color | White |
| Diameter (Metric) Outer | 3.2 mm |
| Diameter (Metric) Inner | 3.2 mm |
| For Use With (Application) | Solvent Handling |
| Packaging Quantity | 5 Fittings |
| For Use With | Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems |
| Material | PFA |
| Autoclavable | Autoclavable |