Learn More
SCAT™ PFA Blind Plug, Type 3
Product Code. 13570810 Shop All SCAT Europe Products
Click to view available options
Thread Size:
1/4-28 UNF
Unit Size:
Pack of 5
Product Code. 13570810
Brand: SCAT™ 160502
Only null left
Add to basket
Add to basket
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
SCAT™ fittings are available in the following versions:
  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications
Type Plug
Color Colorless
For Use With (Application) Solvent Handling
Packaging Quantity 5 Plugs
Thread Size 1/4-28 UNF
For Use With Capillary Connector for HPLC/UHPLC Systems
Material PFA