Learn More
SCAT™ Polypropylene Fitting
Shop All SCAT Europe Products
Click to view available options
Diameter (Metric) Outer:
1.6 mm
2.3 mm
3.2 mm
Unit Size:
5 sheets
Description
SCAT™ fittings are available in the following versions:
- PFA fitting with integrated ferrule in various colors and diameters
- Tube connectors in various diameters
Specifications
Specifications
| Type | Plug |
| Color | Colorless |
| Diameter (Metric) Outer | 1.6 mm |
| For Use With (Application) | Solvent Handling |
| Packaging Quantity | 5 Pk. |
| Thread Size | UNF 1/4 in. 28G |
| For Use With | SCAT Safety Caps |
| Material | Polypropylene |
| Autoclavable | Autoclavable |