Learn More
Memmert™ Flat Stainless Steel Cover
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB11 Water Baths
WTB24 Water Baths
Unit Size:
Each
Description
- Water Bath Accessory
- Stainless Steel
Specifications
Specifications
| For Use With (Equipment) | WTB11 Water Baths |
| Type | Cover |
| Includes | 3 Openings (Ø 87 mm) and Concentric Ring Sets |
| Material | Stainless Steel |
| Shape | Flat |