Learn More
Proteintech™ FlexAble CoraLite™ Plus 550 Antibody Labeling Kit for Mouse IgG1
This Antibody Labeling Kit conjugates CoraLite™ Plus 550 into mouse IgG1 primary antibodies.
Brand:  Proteintech™ KFA022-200
 View more versions of this product
Product Code. 17701529
  • 889.00 EUR / 200 reactions
In Stock
Description
FlexAble CoraLite™ Plus 550 Antibody Labeling Kit for Mouse IgG1 is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 550 in any buffer condition to mouse IgG1 primary antibodies from any supplier.
  • One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
  • Label your mouse IgG1 antibody in <10 min without any additional equipment
Specifications
Specifications
4 x 50 μL CoraLite™ Plus 550 FlexLinker for Mouse IgG1, 4 x 100 μL FlexQuencher for Mouse IgG1, 4 x 500 μL FlexBuffer, Manual, Protocol Magnet
1 Year at -20°C or 6 Months at +4°C, Avoid Light
IgG1
200 x 0.5 μg Reactions
554 nm/570 nm