missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Flotillin 2 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518052
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.
Specifications
| Flotillin 2 | |
| Polyclonal | |
| Unconjugated | |
| FLOT2 | |
| AI573412; ECS1; ECS-1; epidermal surface antigen; ESA; ESA1; FLOT2; flotillin 2; Flotillin 2 (epidermal surface antigen 1); flotillin-2; M17S1; membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); OTTHUMP00000163513; Reg1; REG-1; Reggie-1; reggie1-1; reggie-2 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 2319 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q14254 | |
| FLOT2 | |
| Synthetic peptide sequence (SKIPLIKKATGVQV) corresponding to the C-terminus amino acids of FLOT2 (aa 415-428). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction