missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoxymesterone, Thermo Scientific Chemicals
Fluoxymesterone, CAS # 76-43-7, is a potent, orally administered, synthetic steroid with androgenic activity. It is used in the treatment of male hypogonadism.
591.95 EUR - 2585.87 EUR
Chemical Identifiers
CAS | 76-43-7 |
---|---|
Molecular Formula | C20H29FO3 |
Molecular Weight (g/mol) | 336.447 |
InChI Key | YLRFCQOZQXIBAB-RBZZARIASA-N |
Synonym | fluoxymesterone, fluoximesterone, fluoxymestrone, halotestin, androfluorene, androfluorone, fluotestin, androsterolo, fluosterone, flusteron |
PubChem CID | 6446 |
ChEBI | CHEBI:5120 |
IUPAC Name | (8S,9R,10S,11S,13S,14S,17S)-9-fluoro-11,17-dihydroxy-10,13,17-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one |
SMILES | CC12CCC(=O)C=C1CCC3C2(C(CC4(C3CCC4(C)O)C)O)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10513481
|
Thermo Scientific Acros
124590010 |
1 g |
591.95 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11986881
|
Thermo Scientific Acros
124590050 |
5 g |
2585.87 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Fluoxymesterone is a white, odorless, crystalline powder, soluble in pyridine
- It is an orally-administered synthetic steroid that is about 5-10 times more potent than male steroid, testosterone
- It is metabolized in the liver, and less than 5% of this drug is excreted from the body unchanged
- Adverse effects of fluoxymesterone include sodium and water retention leading to edema, acne, cardiovascular toxicity, and hepatotoxicity
Applications
- Fluoxymesterone has been found to competitively inhibit prolactin or estrogen receptors in tumorigenic breast cells, and is therefore administered in the treatment of breast cancer
- In males, this compound is used as a replacement in conditions associated with symptoms of deficiency or absence of endogenous testosterone
Chemical Identifiers
76-43-7 | |
336.447 | |
fluoxymesterone, fluoximesterone, fluoxymestrone, halotestin, androfluorene, androfluorone, fluotestin, androsterolo, fluosterone, flusteron | |
CHEBI:5120 | |
CC12CCC(=O)C=C1CCC3C2(C(CC4(C3CCC4(C)O)C)O)F |
C20H29FO3 | |
YLRFCQOZQXIBAB-RBZZARIASA-N | |
6446 | |
(8S,9R,10S,11S,13S,14S,17S)-9-fluoro-11,17-dihydroxy-10,13,17-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one |
Specifications
76-43-7 | |
Glass bottle | |
+ 104.00 (20.00°C c=1,EtOH) | |
fluoxymesterone, fluoximesterone, fluoxymestrone, halotestin, androfluorene, androfluorone, fluotestin, androsterolo, fluosterone, flusteron | |
YLRFCQOZQXIBAB-RBZZARIASA-N | |
(8S,9R,10S,11S,13S,14S,17S)-9-fluoro-11,17-dihydroxy-10,13,17-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one | |
6446 | |
336.45 |
240.0°C | |
C20H29FO3 | |
+ 104.00 | |
Solubility in water: negligible. Other solubilities: soluble in puridine, slightly soluble in acetone, chloroform, sparingly soluble in methanol, practically insoluble in ether, benzene, hexanes | |
CC12CCC(=O)C=C1CCC3C2(C(CC4(C3CCC4(C)O)C)O)F | |
336.447 | |
CHEBI:5120 | |
Fluoxymesterone |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Fluoxymesterone
Signal Word
- Warning
Hazard Category
- Reproductive toxicity Category 2
Hazard Statement
- H361d-Suspected of damaging the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title