Learn More
Memmert™ Flush-Fit Unit with Air Slots
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- Stainless steel frame covering gap
- Between oven and wall opening
- Accessory
Specifications
Specifications
| For Use With (Equipment) | HPP110eco |
| Material | Stainless Steel |