missing translation for 'onlineSavingsMsg'
Learn More

FNDC1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA556602

Product Code. 15744511

  • 514.00 EUR / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: DSHPKLSSGI HGDEEDEKPL PATVVNDHVP SSSRQPISRG WEDLRRSPQR GASLHRKEPI PENPKSTGAD THPQGKYSSL A Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 57%.

FNDC1 Polyclonal antibody specifically detects FNDC1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FNDC1
Polyclonal
Unconjugated
FNDC1
1110027O12Rik; 1110051A18Rik; Activation-associated cDNA protein; activator of G-protein signaling 8; AGS8; bA243O10.1; dJ322A24.1; expressed in synovial lining protein; fibronectin type III domain containing 1; fibronectin type III domain containing 2; fibronectin type III domain-containing protein 1; FNDC1; FNDC2; KIAA1866; MEL4B3; mKIAA1866
Rabbit
Antigen affinity chromatography
RUO
84624
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q4ZHG4
FNDC1
Recombinant protein corresponding to Human FNDC1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only