Learn More
Ohaus™ Foam Insert
Product Code. 15712629 Shop All Ohaus Products
Click to view available options
Material:
Foam
Unit Size:
Each
Product Code. 15712629
Brand: Ohaus™ 30400209
Only null left
Add to basket
Add to basket
OHAUS™ Foam Insert is suitable for use with vortex mixers.
  • Offers a large selection of accessories that can be used on all OHAUS vortex mixers
  • Increase throughput by mixing multiple samples at one time
  • Effortlessly mix up to 52 microtubes or eight microplates
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Insert
Material Foam
Color Black
Description Foam Insert Blank