Learn More
Ohaus™ Foam Insert
Shop All Ohaus Products
Click to view available options
Material:
Foam
Unit Size:
Each
Description
- Offers a large selection of accessories that can be used on all OHAUS vortex mixers
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Specifications
| Product Type | Insert |
| Material | Foam |
| Color | Black |
| Description | Foam Insert Blank |