missing translation for 'onlineSavingsMsg'
Learn More

Scharlab FORMALDEHYDLÖSNING 37% PSS1L

Brand:  Scharlab FO00101000


FORMALDEHYDLÖSNING 37% PSS1L

Product Code. 11374124

  • ONLINE EXCLUSIVE PRICE
    52.03 EUR
    46.70 EUR / 1 L
    Save 5.33 EUR
    10% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Formaldehyde, solution 37% w/w, extra pure, Pharmpur, Ph Eur, BP, USP stabilized with approx. 10%

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H311 + H331-Toxic in contact with skin or if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde, Methanol

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.