Learn More
Watson-Marlow 314DW Pumphead
Click to view available options
For Use With:
323 Series Peristaltic Pumps
Unit Size:
Each
Description
- The 300 series combines performance, practicality and design. With its iconic flip-top design it is an industry standard OEM product in medical device, biotechnology and diagnostic applications, and very popular in many other applications too.
- Case material: Glass filled polypropylene, IXEF (Polyarylamide), and Stainless steel
- Rotor material: Electroless nickel plated hardened steel, and Glass filled Nylon
Specifications
Specifications
| For Use With | 323 Series Peristaltic Pumps |
| No. of Rollers | 4 |
| Type | Pump Head |