missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO1/FOXO3/FOXO4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538274
This item is not returnable.
View return policy
Description
FOXO1 + FOX3 + FOX4 belong to the forkhead family of transcription factors. FOXO1 (FKHR or ForkHead in Rhabdomyosarcoma) is a 70 kDa protein. Recent studies have shown that this protein can act as either a coactivator or a corepressor of nuclear receptor activity. This action is mediated through the LXXLL motif found in the C terminus of the FOXO1A protein. The specific function of the gene has not yet been determined.FOX3 probably functions as a trigger for apoptosis through expression of genes necessary for cell death. Translocation of the FOXO3 gene with the MLL gene is associated with secondary acute leukemia.FOX4 is a forkhead transcription factor involved in the regulation of the insulin signaling pathway. It binds to insulin-response elements (IREs) and can activate transcription of IGFBP1. FOXO4 down-regulates expression of HIF1A and suppresses hypoxia-induced transcriptional activation of HIF1A-modulated genes. It is also involved in negative regulation of the cell cycle.Specifications
FOXO1/FOXO3/FOXO4 | |
Polyclonal | |
Unconjugated | |
FOXO4 | |
Forkhead box protein O4, Fork head domain transcription factor AFX1, AFX, AFX1, MLLT7, FOXO4, Forkhead box protein O1, Forkhead box protein O1A, Forkhead in rhabdomyosarcoma, FOXO1, FKHR, FOXO1A, Forkhead box protein O3, AF6q21 protein, Forkhead in rhabdomyosarcoma-like 1, FOXO3, FKHRL1, FOXO3A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2308, 2309, 4303, 54601, 56458, 56484 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43524, P98177, Q12778, Q9R1E0, Q9WVH3, Q9WVH4 | |
FOXO1, FOXO3, FOXO4 | |
A synthetic peptide derived from the internal region of human FOXO1/3/4-pan | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |