Learn More
Brady™ FreezerBondz™ Cryogenic Matte Polyester Laboratory Labels for M6/M7 Printers
Durable Polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 173708
 View more versions of this product
Product Code. 17878144
  • 55.30 EUR / Pack of 100
In Stock
to see stock.
Explore more special offers
Description
  • B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen when wrapped around itself by at least 1/8 in.
  • Good print smudge resistance, solvent resistance and high/low temperature performance of -196°C to 100°C (-320°F to 212°F)
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Specifications
Specifications
Autoclavable
M6-113-490
None
Matte
Polyester
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
100 Labels
-320°F to +266°F
0.0026 in.
2.2 x 1.5 in.
1.5 in.
55.88 mm
ASTM D 1000, ASTM G 155
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
Laboratory Labeling
No
Rectangle
-196°C to +130°C
0.065 mm
2.2 in.
38.1 mm