Learn More
MULTIROIR Full Eurobox Container
Click to view available options
Dimensions (L x W x H):
400 x 300 x 117 mm
400 x 300 x 120 mm
400 x 300 x 220 mm
400 x 300 x 290 mm
600 x 400 x 100 mm
600 x 400 x 220 mm
600 x 400 x 320 mm
600 x 400 x 420 mm
Color:
Blue
Gray
Red
White
Unit Size:
Each
Description
- Full Eurobox container
- The best quality / price ratio on the market
- Solid bottoms and solid walls
- Ideal for the storage and transport of goods
- Stackable and high load capacity
- Temperature resistant from -20 ° C to + 110 ° C
Specifications
Specifications
| Type | Storage Box |
| Material | Polypropylene |
| Color | White |
| Shape | Rectangular |
| Dimensions (L x W x H) | 600 x 400 x 220 mm |