Learn More
Memmert™ Full-Sight Glass Door for UF1060plus, Max. 300°C
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF1060plus Oven
Unit Size:
Each
Description
- Accessory
- Full-sight
- Glass (4 layers)
- Temperature range up to 300°C
Specifications
Specifications
| For Use With (Equipment) | UF1060plus Oven |
| Temperature (Metric) | 300°C (Max.) |
| Type | Full-sight Door |
| Material | Glass |