Learn More
Memmert™ Full-Sight Glass Door for UF750plus, Max. 300°C
Product Code. 17569575 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF750plus Oven
Unit Size:
Each
Product Code. 17569575
Brand: Memmert™ UF750plusB1
Only null left
Add to basket
Add to basket
Full-sight glass door constructed of four insulated glass layers.
  • Accessory
  • Full-sight
  • Glass (4 layers)
  • Temperature range up to 300°C
Specifications
For Use With (Equipment) UF750plus Oven
Temperature (Metric) 300°C (Max.)
Type Full-sight Door
Material Glass