missing translation for 'onlineSavingsMsg'
Learn More

Kimble™ Glass Funnel

Measure and filter substances in the laboratory with the Kimble™ Glass Funnel, an essential experimentation tool.

Brand:  Kimble™ 953701-0000

Product Code. 11858522

  • 146.00 EUR / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Weight: 0.8kg
  • Clear glass
  • Graduated for accurate measurement
TRUSTED_SUSTAINABILITY
Specifications

Specifications

25mm
15 mL
Glass
Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.