Learn More
Funnel Stand Replacement Parts
Click to view available options
For Use With (Equipment):
Spare for STA-200-061L
Spare for STA-200-071Y
Dimensions (L x W):
205 x 110mm
290 x 110mm
Unit Size:
Each
Description
Hardwood
Specifications
Specifications
| Includes | With plastic screw and Rubber feet on base |
| Product Type | Funnel Stand |
| Holds | Single |
| Material | Hardwood |
| For Use With (Equipment) | Spare for STA-200-061L |
| Size | 205 x 110 mm |
| Dimensions (L x W) | 205 x 110mm |
| Length (Metric) | 460 mm |