missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl mercaptan, 97%, Thermo Scientific Chemicals

Product Code. 10377080
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-02-2
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00003254
InChI Key ZFFTZDQKIXPDAF-UHFFFAOYSA-N
Synonym furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol
PubChem CID 7363
IUPAC Name furan-2-ylmethanethiol
SMILES C1=COC(=C1)CS

Specifications

Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 155.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5290 to 1.5330
Quantity 100 mL
Beilstein 17, II, 116
Specific Gravity 1.13
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in chloroform
Formula Weight 114.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Furfuryl mercaptan, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Furfuryl mercaptan
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.