Learn More
Carbolite™ Furnace with Uniform Temperature Envelope
Product Code. 12933741
Click to view available options
Capacity (Metric):
5 L
15 L
23 L
Temperature (Metric):
1200°C
1300°C
1600°C
Dimensions (D x W x H) Interior:
250 x 140 x 135 mm
305 x 220 x 220 mm
400 x 245 x 235 mm
Unit Size:
Each
Product Code. 12933741
Brand: Carbolite™ CWF1305230SN+&02301
Only null left
Specifications
Controller Type 301 PID Controller
Capacity (Metric) 5 L
Temperature (Metric) 1300°C
Type Furnace
Dimensions (D x W x H) Interior 250 x 140 x 135 mm
Width (Metric) 375 mm
Width (Metric) Interior 140 mm
Height (Metric) 585 mm
Height (Metric) Interior 135 mm
Depth (Metric) 485 mm
Depth (Metric) Interior 250 mm
Electrical Requirements 230 V 50/60 Hz 2400 w
Voltage 230 V
Wattage 2400 w
Frequency 50/60 Hz