Learn More
G-CSFR/CD114 Antibody (723806), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB60391APCCY7
Product Code. 30039749
  • 644.00 EUR / 0.10mL
Explore more special offers
Description
Description
G-CSFR/CD114 Monoclonal antibody specifically detects G-CSFR/CD114 in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
G-CSFR/CD114
Monoclonal
APC-Cyanine7
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
1441
Store at 4C in the dark. Do not freeze.
IgG2a
Flow Cytometry
723806
Flow Cytometry
CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), G-CSF receptor, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor
Mouse myeloma cell line NS0-derived recombinant mouse G-CSF R/CD114, Cys26-Asp626, Accession # P40223
0.1 mL
Primary
Mouse
Purified