Learn More
Emma Safety Footwear Galus S3 Duo Bond Boot
Product Code. 17562476
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17562476
Brand: Emma Safety Footwear MM58086646
Only null left
Add to basket
Add to basket
Insulated with fine quality sheepskin, there is no warmer shoe than the Galus to wear in the coldest of conditions.
  • A naturally better boot.
  • Even in this age of hi-tec, high performance synthetics, sometimes nature's materials are still the best. Which is why we've insulated the Galus with fine quality sheepskin. There's no warmer shoe to wear in the coldest of conditions. And if the temperature suddenly rises up to 300 degrees on the outside, it's reassuring to know the nitrile rubber outsole won't yield one bit.
  • Industries:
    • Chemical industry
    • Construction & technology
  • Safety shoe
    • S3-Safetyshoe with sole plate for wet work
Specifications
Certifications/Compliance ISO 20345:2011
Closure Type Zipper
Color Black
For Use With (Application) Chemical Industry, Construction and Technology
Liner Material Sheepskin
Material Leather
Size 46
Sole Features GripForce™ Duo
Sole Material Nitrile Rubber
Toe Material Steel
Fit Extra-wide Width
EN ISO 20345 S3
Product Type Safety Boot