Learn More
Keystone Cleanroom Products Gamma-Irradiated 2 mil Polyethylene Trash Bag
Shop All Keystone Cleanroom Products Products
Click to view available options
Includes:
1 Pack
10 Packs
2 Packs
5 Packs
Unit Size:
Pack of 10
Pack of 100
Pack of 40
Pack of 50
Description
- Packaged in an ISO6 cleanroom
- 2 mil clear polyethylene
- Each package includes a lot number for traceability
- Certificates of Conformance and Irradiation sent with shipment
- Packaged in Tyvek™/Mylar Chevron Peel Pouches
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 16.3 L |
| Color | Clear |
| Includes | 2 Packs |
| Material | LDPE |
| Thickness (Metric) | 0.05 mm |
| Thickness (English) | 2 mil |
| Dimensions (L x W) | 71 x 61 cm (28 x 24 in.) |