Learn More
Cytiva GammaBind™ Plus Sepharose™ Medium
Product Code. 11504955 Shop All Cytiva Products
Click to view available options
Quantity:
5 mL
25 mL
500 mL
Unit Size:
25mL
500mL
5mL
Product Code. 11504955
Brand: Cytiva 17088604
Engineered for better binding to rat and mouse IgG subclasses
  • Recombinant Protein G type 3 coupled to Sepharose CL-6B for easy binding and fast separation
  • More enhanced binding capabilities than GammaBind G at both laboratory and process scale
  • Yields up to 18mg/mL mouse IgG
  • Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
TRUSTED_SUSTAINABILITY
Specifications
Product Type GammaBind Plus Sepharose
Ligand Recombinant GammaBind G type 3 lacking albumin-binding region
Particle Size 90 μm
Content And Storage 2°C to 8°C, 20% Ethanol
Quantity 500 mL
compliance-icons
Product Identifier
  • GammaBind Plus Sepharose, 500 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol