missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Schmidlin™ GC Plus Zero Air Generator
This generator produces a continuous supply of zero grade air.
Brand: Schmidlin™ DB-NGC15000-EU
This item is not returnable.
View return policy
Description
- Increase efficiency and improve safety and performance
- On-demand supply 24/7
- Purity: <0.1 ppm THC
- Pressure: 10 barg (145 psig)
- Proprietary platinum catalyst technology
- Easy to install with complete 'plug and play' functionality
- Ideal for all GC detector applications
- Stable baseline with increased sensitivity and repeatability
- Meets and exceeds requirements for the most demanding GC applications
Specifications
23 cm | |
41 cm | |
3.5 to 9 bar | |
55 cm | |
110/120 V at 60 Hz, 220/240 V at 50 Hz | |
550 W |
15,000 mL/min. | |
35°C | |
21.6 in. | |
Inlet: 1/4 in., Outlet: 1/8 in. | |
<0.1 ppm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction