missing translation for 'onlineSavingsMsg'
Learn More

Schmidlin™ GC Plus Zero Air Generator

This generator produces a continuous supply of zero grade air.

Brand:  Schmidlin™ DB-NGC30000-EU

Encompass_Preferred

 View more versions of this product

Product Code. 17728909

  • 13166.62 EUR / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Purity: <0.1 ppm THC
  • Pressure: 10 barg (145 psig)
  • Proprietary platinum catalyst technology
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC detector applications
  • Stable baseline with increased sensitivity and repeatability
  • Meets and exceeds requirements for the most demanding GC applications
Specifications

Specifications

23 cm
41 cm
3.5 to 9 bar
55 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
550 W
30,000 mL/min.
35°C
21.6 in.
Inlet: 1/4 in., Outlet: 1/8 in.
<0.1 ppm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers