Learn More
Thermo Scientific™ GeneBLAzer™ D5-CRE-bla CHO-K1 Cells
Description
The GeneBLAzer™ D5-CRE-bla CHO-K1 cells contain the human Dopamine Receptor 5 (D5) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase reporter gene under control of the CRE response element.
The GeneBLAzer™ D5-CRE-bla CHO-K1 cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, GeneBLAzer™ D5-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.
Specifications
Specifications
| Accession Number | NM_000676 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ D5-CRE-bla CHO-K1 cells • GeneBLAzer™ D5-CRE-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ D5-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Detection Method | Fluorescence |
| For Use With (Application) | GPCRs |
| Gene Symbol | DRD5 |
| Includes | D5-CRE-bla CHO-K1 cells, D5-CRE-bla CHO-K1 Assay Protocol, Certificate of Analysis |
| Population Status | Single Cell Clone Origin |
| Product Line | GeneBLAzer |
| Show More |