Learn More
Thermo Scientific™ GeneBLAzer™ GR-UAS-bla HEK 293T Cells
GR-UAS-bla HEK 293T cells contain the ligandbinding domain (LBD) of the human Gluocorticoid receptor (GR) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line.
Brand:  Thermo Scientific™ K1687
Product Code. 10083913
  • 10380.00 EUR / Each
Estimated Shipment: 20-01-2025
Description
GR-UAS-bla HEK 293T cells contain the ligandbinding domain (LBD) of the human Gluocorticoid receptor (GR) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-blaHEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-GR (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. GR-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of Dexamethasone. In addition, GR-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tetrac, T4
Cell-based beta lactamase reporter gene
GR-UAS-bla HEK 293T cells (K1687)
Shipping: Dry Ice
Storage: -80°C (liquid nitrogen), Liquid Nitrogen
GeneBLAzer™ GR-UAS-bla HEK 293T Cells, 2 x 106 Cells, ∽2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, 1-850 Antagonists, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Human
GeneBLAzer™ GR-UAS-bla HEK 293T cells - Includes: GR-UAS-bla HEK 293T cells, Protocol, Certificate of Analysis
T3 Hormone
2 x 106 Cells, 2 x 106 cells
293T (HEK)
NR3C1, GR
1-850
Dividing Cells
Fluorescence, Fluorescent
Pharma and biopharma, target and lead identification and validation
Nuclear Receptors
2 x 106 Cells
GeneBLAzer™
HEK 293T Cells
BLA (Beta-Lactamase)
Cell Line