missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GeneBLAzer™ PAC1 CHO-K1 DA Assay Kit

GeneBLAzer™ PAC1 CHO-K1 DA Assay Kit

Brand:  Invitrogen™ K1619

Product Code. 10791614

  • 966.00 EUR / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells contain the human Adenylate Cyclase Activating Polypeptide 1 Receptor 1 (ADCYAP1R⁄PAC1) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat #K1129) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element (CRE). The PAC1-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of established ligands. In addition, PAC1-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary. Division Arrested cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM_001118
Division-Arrested Cells
Fluorescence
Single Cell Clone Origin
8 x 106 cells
ADCYAP1R1, PAC1
CHO-K1
GPCR
Cell-based beta lactamase reporter gene
K1619 GeneBLAzer™ PAC1 CHO-K1 DA Assay Kit
Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ PAC1 CHO-K1 DA cells
• LiveBLAzer™-FRET B⁄G Loading Kit, 70μg
• Solution D
• GeneBLAzer™ PAC1 CHO-K1 DA Assay Protocol
• LiveBLAzer™-FRET B⁄G Loading Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer™ PAC1 CHO-K1 DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
ADCYAP1R1
GeneBLAzer™
BLA (Beta-Lactamase)
Division-Arrested Cells Assay Kit
GeneBLAzer™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.