missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ GeneClean II Kit

GeneClean™ II Kit

Brand:  MP Biomedicals™ 111001400


Product Code. 11383099

  • 380.00 EUR / 300 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The GENECLEAN™ II Kit is designed to optimally purify 200 bp to 20 kb DNA from TAE and TBE buffered agarose gels and solutions. Glassmilk™ slurry allows flexibility of reacion size and volume.
  • The purified DNA is suitable for a variety of downstream applications, such as sequencing, PCR, restriction digestion, cloning and labeling.
  • The GENECLEAN™ II Kit is designed for the rapid purification of linear and plasmid DNA fragments from standard or low-melting agarose gels.
  • This kit can also be used to purify DNA from polyacrylamide gels.
  • Typical recovery of DNA is up to 80%.
  • Each column can bind up to 10 μg of DNA from up to a 3.5 g agarose slice.
  • The GENECLEAN™ II Kit combines silica-binding technology with the convenience of a spin or vacuum column format.
  • DNA fragments of interest are extracted from slices of an agarose gel and are bound to a silica membrane.
  • Contaminants are removed by a simple spin or vacuum wash.
  • The bound DNA is then eluted.
  • DNA fragments of interest are extracted by solubilizing slices of an agarose gel.
  • The Gel Solubilization Solution dissolves an agarose gel slice from gels run in TBE or TAE buffer.
  • The extracted DNA fragments selectively adsorb onto the silica membrane in the presence of the Gel Solubilization Solution.
  • Contaminants are removed by a simple spin or vacuum wash. Finally, the bound DNA is eluted in Tris buffer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

New Wash/Ethanol Concentration must be ∽ 50%
Nucleic Acid Isolation & Purification
Above 500 bp: 4.5 M NaI pH 7.0 - 7.4; Between 200 - 500 bp: 4.5 M NaI pH 6.0; NEW Wash: pH Range: 7.4 - 8.0
Rapid purification of linear DNA
All gENECLEAN™ Kits are shipped at room temperature (15-30°C).
300 Preps
Kits & Assays
Solution or Gels
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • geneclean II kit

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.

Supplemental information
  • MIXTURE LIST-Contains : silicon dioxide, acetic acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers