Learn More
Brannan™ Lab Thermometer with 3-points UKAS Calibration
Product Code. 17823088
Click to view available options
Temperature Range (Metric):
-10°C to +110°C
-10°C to +150°C
-10°C to +50°C
-100°C to +50°C
-18°C to +110°C
-20°C to +110°C
-20°C to +150°C
Thermometer Fill:
Green Spirit
Red Spirit
Spirit
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
15 cm
20.5 cm
30.5 cm
Unit Size:
Each
Product Code. 17823088
Brand: Brannan™ 46/456/0 + 49/503/0
Only null left
Add to basket
Add to basket
General purpose laboratory thermometer complete with calibration certificate.
  • Lab thermometer
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
Calibration Points -60°C, 0°C, 40°C
Certifications/Compliance ISO 17025 UKAS
Immersion Type Total Immersion
Length (Metric) 30.5 cm
Material Glass
Thermometer Fill Red Spirit
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) -148°F
Min. Temperature (Metric) -100°C
Temperature Range (English) -148°F to +122°F
Temperature Range (Metric) -100°C to +50°C
Accuracy ± 2.0°C at 35°C
Subdivision 1°C
Width (Metric) 1.2 cm