Learn More
3M™ General Purpose Masking Tape 201E
Click to view available options
Dimensions (LxW):
50 m x 24 mm
50 m x 48 mm
Unit Size:
Pack of 24
Pack of 36
Descripción
- This tape is designed for general use in industrial applications with a slight need for temperature and water resistance such as short-term paint masking and sealing applications.
- It provides a good instant adhesion and holding power that resists lifting or curling.
- Its crepe paper backing offers conformability that will maintain integrity when formed around corners.
- Its good solvent and moisture resistance helps to resist paint bleed through and provides good paint lines due to its clean removability in one piece with no residue from most surfaces.
Especificaciones
Especificaciones
| Color | Beige |
| Material | Crepe Paper |
| Product Type | Masking Tape |
| Dimensions (LxW) | 50 m x 48 mm |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido