missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitlab™ Genius Bottle Top Dispensers
Equipped with a positive displacement piston and a PFA sealing lip on the cylinder wall. Vitlab™ Genius Bottle Top Dispensers, with proven precision, offer many advantages in routine liquid-handling operations.
Brand: Vitlab™ 1605505
This item is not returnable.
View return policy
Description
- Materials that come into in contact with media (PTFE, PFA, FEP, borosilicate glass and platinum-iridium) are resistant to most acids, bases, and solvents
- Reagent loss while ventilating is avoided with the innovative recirculation valve
- Simple-to-use calibration function helps meet all the requirements for test equipment monitoring without downtime
- Sealing lip acts like a windscreen wiper to prevent crystal build-up on the cylinder wall from readily crystallisable media
- Glass cylinder is coated with a plastic material that reduces the risk of splashes should breakage occur
- Telescopic filling tube can be adjusted smoothly to different bottle heights
- Autoclavable at 121 °C (2 bar) according to DIN EN 285
- DE-M marked
- Included in delivery:
- Dispenser with 3 threaded adapters made from PP
- Nominal volumes of 2.5 – 10 ml (screw coupling GL 32) with adapters GL 28, S 40 and GL 45
- Nominal volumes of 25 – 100 ml (screw coupling GL 45) with adapters GL 32, GL 38 and S 40
- Telescopic filling tube (200 – 350 mm), mounting tool, instruction manual, and quality certificate stating all test values.
Specifications
±0.5% | |
Autoclavable | |
1 to 10 mL | |
15 Months |
Distributor | |
DIN EN ISO 8655-3 | |
0.2 mL |