missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ G-CSF Recombinant Mouse Protein
Recombinant G-CSF is a bioactive protein intended for use in cell culture applications
1656.00 EUR
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 19 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | G-CSF |
Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10151243
|
Gibco™
PMC2031 |
100μg | |||||||
Description
G-CSF stimulates the proliferation and differentiation of hematopoietic progenitor cells.Specifications
Cell Culture | |
Sequential Chromatography | |
Mouse | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
GM-CSF, G-CSF, & M-CSF | |
Gibco™ |
19 kDa | |
G-CSF | |
Recombinant | |
Carrier-Free | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Signal Transduction, Oncology, Stem Cell Research, Differentiation, Drug Development | |
95 % |
For Research Use Only. Not for use in diagnostic procedures.