Learn More
Cytiva Glass Plates for Standard Flatbed Units
Supplied in packs of 3
Brand: Cytiva 2117-932
This item is not returnable.
View return policy
Description
- 125 x 240 x 0.5 mm
- Polystyrene spacer
Specifications
Standard Flatbed Unit | |
Glass | |
3.0 mm | |
260 mm |
U-Frame Glass Plate | |
0.11 in. | |
125 mm |