Learn More
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispenser
Product Code. 10558721
Click to view available options
Volume (Metric):
0.4 to 2 mL
1 to 5 mL
10 to 50 mL
2 to 10 mL
20 to 100 mL
40 to 200 mL
6 to 30 mL
Unit Size:
Each
Product Code. 10558721
Brand: Poulten Graf™ 101 08144
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
For dispensing of pure organic solvents. Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • Solid dispensing piston made of glass
  • Cylinder coated with plastic to protect against breakage
  • With certificate, individually numbered
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Product Type Fortuna Optifix Solvent Dispenser
Channels 1
Material PTFE, Polypropylene, Fiberglass
Volume (Metric) 6 to 30 mL
For Use With (Application) For dispensing solvents
Quantity Each