Learn More
Hirschmann™ Hollow Glass Stoppers
Click to view available options
Standard Taper Joint:
NS 10/19
NS 12/21
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Description
- Acc. to DIN 12 252
- Available to fit a range of sizes
Specifications
Specifications
| For Use With (Equipment) | For tight sealing of standard ground vessels |
| Quantity | 1/Ea. |
| Standard Taper Joint | NS 12/21 |
| Material | Glass |
| Product Type | Hollow Stopper |