Learn More
Poulten Graf™ Glass Volac Burette
Brand:  Poulten Graf™ RB9803/WAC/Z
Product Code. 10331412
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Order Info
dimCapacityMetric
Specifications
Specifications
10 mL
0.05 mL
Straight
Volumetric Buret
Glass
Blue
PTFE
Class AS