missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Glove

Product Code. 12693373
Click to view available options
Size: 8
This item is not returnable. View return policy
This item is not returnable. View return policy

Protect your hands from very hot objects with these ambidextrous, knitted cotton gloves covered in resistant nitrile dots. Honeywell Safety Products™ Aratherma Fit Short Grip Gloves absorb hand perspiration and provide both cutting and thermal resistance.

  • 100% knitted cotton glove provides hand conformity, dexterity, cutting resistance and contact heat resistance.
  • Double face coating of nitrile dots allows for handling of very hot objects with comfort and grip.
  • Ambidextrous.
  • Cotton lining reinforces mechanical performance and absorbs hand perspiration.
  • High quality materials create an effective barrier against heavy and sharp parts.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standard, EN 388-2003 Mechanical hazards, EN 407-2004 Thermal hazards, EC Category PPE 2, EC Certificate Number 0075/797/162/09/08/0328, EC Attestation Number 0075/797/162/09/08/0328
Classification Dexterity: 3
Coated Area Palm, Fingers
Construction String Knit
Description Gauge 10
Coating Material Nitrile Dots
Cut Resistance Rating 2
Hand Ambidextrous
Material Nitrile
Product Type Gloves
Size 8
For Use With (Application) Handling of very hot objects. Automotive, Glass, Iron and Steel industries.
Quantity 6Pair/PK
Resistant To Heat
Tear Rating 3
Length (Metric) 25 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'